Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL17F Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
209-401-B31
RL209-401-B31EA 518 USD InStock
RL209-401-B31
Anti-IL17F Rabbit Polyclonal Antibody
Antibodies
This purified antibody has been tested for use in neutralizations, ELISA and western blotting. Reactivity is also expected in radioimmunoassay.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: IL17F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty