Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GDF15 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
209-401-B88 209-401-B88S
RL209-401-B88SEA 148 USD InStock
RL209-401-B88S RL209-401-B88
Anti-GDF15 Rabbit Polyclonal Antibody
Antibodies
This Protein-A purified, MBP-cross-adsorbed antibody is suitable for ELISA and western blotting of human and mouse NAG-1 Protein.   For detection of NAG-1 in human serum, a sandwich ELISA is suggested using this antibody in combination with anti-NAG-1/GDF.

Recommended Dilutions: ELISA: 1:500; Western Blot: 1:800 - 1:1500;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty