Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL20 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
209-403-B51
RL209-403-B51EA 498 USD InStock
RL209-403-B51
Anti-CCL20 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
This purified antibody has been tested for use in ELISA and western blotting. By western blot a band approximately 11 kDa in size corresponding to human MIP-3a Protein is expected in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:500 - 1:5000;
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Primary
Antigen: CCL20
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty