Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BIN1 Mouse Monoclonal Antibody [clone: 12A]

Supplier: Rockland Immunochemicals
209-301-E62S
RL209301E62SEA 148 USD InStock
RL209301E62S
Anti-BIN1 Mouse Monoclonal Antibody [clone: 12A]
Antibodies
Type: Primary
Antigen: BIN1
Clonality: Monoclonal
Clone: 12A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty