Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNF Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
210-401-321 210-401-321
RL210-401-321EA 1290 USD InStock
RL210-401-321 CA210-401-321
Anti-TNF Rabbit Polyclonal Antibody
Antibodies
This antibody against Mouse TNFa has been tested for use in neutralizations, ELISA, immunochemistry, radioimmunoassay, immunoprecipitation, immunochemistry and immunoblotting.  It recognizes the 17,000 MW TNFa.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: TNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty