Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL17A Rat Monoclonal Antibody [clone: 20B4.G10.F5]

Supplier: Rockland Immunochemicals
210-501-B32 210-501-B32S
RL210-501-B32EA 528 USD InStock
RL210-501-B32 RL210-501-B32S
Anti-IL17A Rat Monoclonal Antibody [clone: 20B4.G10.F5]
Antibodies
IL-17A monoclonal antibody has been tested for use in western blotting, flow cytometry and ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 23 kDa in size.

Recommended Dilutions: ELISA: 1:10000; Flow Cytometry: 1:500; Western Blot: 1:1000;

Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: 20B4.G10.F5
Conjugation: Unconjugated
Epitope: Full length
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty