Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL20 Rat Monoclonal Antibody [clone: 4N5F7]

Supplier: Rockland Immunochemicals
210-501-B51S
RL210-501-B51SEA 148 USD InStock
RL210-501-B51S
Anti-CCL20 Rat Monoclonal Antibody [clone: 4N5F7]
Antibodies
Type: Primary
Antigen: CCL20
Clonality: Monoclonal
Clone: 4N5F7
Conjugation: Unconjugated
Epitope: Full length
Host: Rat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty