Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL27 Rat Monoclonal Antibody [clone: 3H12.F10]

Supplier: Rockland Immunochemicals
210-501-B54 210-501-B54S
RL210-501-B54EA 528 USD InStock
RL210-501-B54 RL210-501-B54S
Anti-IL27 Rat Monoclonal Antibody [clone: 3H12.F10]
Antibodies
IL-27 is expressed in activated antigen presenting cells including monocytes, endothelial cells, and dendritic cells, for example mouse CD4 splenocytes. This purified antibody has been tested for use in Flow Cytometry, ELISA and western blotting.

Recommended Dilutions: ELISA: 1:10000; Flow Cytometry: 1 µg/ml; Western Blot: 1:1000;

Type: Primary
Antigen: IL27
Clonality: Monoclonal
Clone: 3H12.F10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty