Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EBI3 Rat Monoclonal Antibody [clone: 10J811]

Supplier: Rockland Immunochemicals
210-501-B66S
RL210-501-B66SEA 148 USD InStock
RL210-501-B66S
Anti-EBI3 Rat Monoclonal Antibody [clone: 10J811]
Antibodies
Type: Primary
Antigen: EBI3
Clonality: Monoclonal
Clone: 10J811
Conjugation: Unconjugated
Epitope: Full length
Host: Rat
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty