Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IDO2 Mouse Monoclonal Antibody [clone: 1HC]

Supplier: Rockland Immunochemicals
210-301-E59S
RL210301E59SEA 148 USD InStock
RL210301E59S
Anti-IDO2 Mouse Monoclonal Antibody [clone: 1HC]
Antibodies
Type: Primary
Antigen: IDO2
Clonality: Monoclonal
Clone: 1HC
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty