Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (Agarose)

Supplier: Rockland Immunochemicals
211-1150 211-1150
RL211-1150EA 329 USD InStock
RL211-1150 CA211-1150
Anti-IgG Goat Polyclonal Antibody (Agarose)
Antibodies
Secondary Goat Anti-IgG (H&L) Reacts with Rabbit (Lapine)

Recommended Dilutions:
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Agarose
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty