Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Rat Serum Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
212-4301-0100 212-4301-0100
RL21243010100EA 488 USD InStock
RL21243010100 CA10037-230
Anti-Rat Serum Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: Rat Serum
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty