Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RBC Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
213-4139 213-4139
RL213-4139EA 434 USD InStock
CA213-4139 RL213-4139
Anti-RBC Rabbit Polyclonal Antibody
Antibodies
Suitable for agglutination of cells on titer plates. Each laboratory should determine an optimum working titer for use in its particular application. Other applications have not been tested but use in such assays should not necessarily be excluded.

Recommended Dilutions: Agglutination Titer 1:32 - 1:64;
Restore with deionized water (or equivalent); 5.0 ml

Type: Primary
Antigen: RBC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Sheep

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty