Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgM µ Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Rockland Immunochemicals
213-4207 213-4207
RL213-4207EA 549 USD InStock
CA213-4207 RL213-4207
Anti-IgM µ Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Secondary Rabbit Anti-IgM mu chain Reacts with Sheep (Ovine)

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: µ-chain
Host: Rabbit
Isotype: IgM
Reactivity: Sheep

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty