Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNF Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
214-401-306S 214-401-306
RL214-401-306EA 518 USD InStock
RL214-401-306 RL214-401-306S
Anti-TNF Rabbit Polyclonal Antibody
Antibodies
This Protein A purified antibody is suitable for ELISA and Western blotting.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 16.9 kDa in size corresponding to swine TNFa by western blotting.

Recommended Dilutions: ELISA: 1:2000 - 1:2500; Western Blot: 1:1000;

Type: Primary
Antigen: TNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty