Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Pancreatic islet lysate Dog Polyclonal Antibody

Supplier: Rockland Immunochemicals
214-A01-118 214-A01-118
RL214-A01-118EA 570 USD InStock
RL214-A01-118 CA214-A01-118
Anti-Pancreatic islet lysate Dog Polyclonal Antibody
Antibodies
This product has been assayed against 1.0 µg of Pancreatic Islet Lysate (Porcine Pancreas) in a standard sandwich ELISA using Peroxidase conjugated Affinity Purified anti-Dog IgG (H&L) (Goat) and o-Phenylenediamine (OPD) as a substrate.

Recommended Dilutions:

Type: Primary
Antigen: Pancreatic islet lysate
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Dog
Isotype: IgG
Reactivity: Pig

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty