Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MTBP Mouse Monoclonal Antibody [clone: PHL-1]

Supplier: Rockland Immunochemicals
500-301-875 500-301-875
RL500-301-875EA 528 USD InStock
RL500-301-875 CARL500301-875
Anti-MTBP Mouse Monoclonal Antibody [clone: PHL-1]
Antibodies
This antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band ~ 110 kDa in size corresponding to MTBP by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:2000 - 1:20000; Western Blot: 1:200 - 1:2000;

Type: Primary
Antigen: MTBP
Clonality: Monoclonal
Clone: PHL-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty