Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-H5N1 Mouse Monoclonal Antibody [clone: 7A11]

Supplier: Rockland Immunochemicals
500-301-977
RL500-301-977EA 528 USD InStock
RL500-301-977
Anti-H5N1 Mouse Monoclonal Antibody [clone: 7A11]
Antibodies
This monoclonal antibody can be used for hemagglutination inhibition (HI) assays to provide antigenic characterization of the influenza A viruses of the H5 HA subtype.

Recommended Dilutions: ELISA: 1:5000;

Type: Primary
Antigen: Hemagglutinin 5
Clonality: Monoclonal
Clone: 7A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: H5N1 virus, Virus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty