Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KDELR1 Mouse Monoclonal Antibody [clone: KR-10]

Supplier: Rockland Immunochemicals
500-301-F86
RL500-301-F86EA 532 USD InStock
RL500-301-F86
Anti-KDELR1 Mouse Monoclonal Antibody [clone: KR-10]
Antibodies
Anti-KDEL Antibody is suitable for WB, IP, Immunofluorescence microscopy and IHC. Expect a band approximately ~25kDa Protein corresponding to the molecular mass of KDEL on SDS-PAGE immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: KDELR1
Clonality: Monoclonal
Clone: KR-10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Rat, Mouse, Hamster, Rabbit, Pig, Sheep, Dog, Chicken, Drosophilia, Xenopus, Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty