Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Fluorescein Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanante))

Supplier: Rockland Immunochemicals
600-100-096 600-100-096
RL600-100-096EA 697 USD InStock
RL600-100-096 CA600-100-096
Anti-Fluorescein Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanante))
Antibodies
Anti-Fluorescein Rhodamine Conjugated Antibody is suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: 1:500 - 1:2500; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: Fluorescein
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanante)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty