Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ING4 Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-101-284 600-101-284
RL600-101-284EA 518 USD InStock
RL600-101-284 CA600-101-284
Anti-ING4 Goat Polyclonal Antibody
Antibodies
This affinity-purified antibody has been tested for use in ELISA against the immunizing peptide.  Specific conditions for western blotting reactivity should be optimized by the end user. Expect a band at approximately 29 kDa in size corresponding to ING4.

Recommended Dilutions: ELISA: 1:3000 - 1:12000; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: ING4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty