Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Fluorescein Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
600-103-096 600-103-096
RL600-103-096EA 678 USD InStock
RL600-103-096 CA600-103-096
Anti-Fluorescein Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Fluorescein Antibody Peroxidase Conjugated Suitable for immunoblotting (western or dot blot), ELISA, and in situ hybridization, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays

Recommended Dilutions: ELISA: 1:200,000; Western Blot: 1:5000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Tag
Antigen: Fluorescein
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty