Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RPE Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemicals
600-105-387
RL600-105-387EA 744 USD InStock
RL600-105-387
Anti-RPE Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Suitable for immunoblotting (western or dot blot), ELISA and immunochemistry as well as other phosphatase-antibody based enzymatic assays requiring lot-to-lot consistency.  R-Phycoerythrin (240 kDa) is a labile molecule that may dissociate into compo

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000
Do not freeze!

Type: Tag
Antigen: RPE
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty