Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Fluorescein Goat Polyclonal Antibody (DyLight® 800)

Supplier: Rockland Immunochemicals
600-145-096
RL600-145-096EA 487 USD InStock
RL600-145-096
Anti-Fluorescein Goat Polyclonal Antibody (DyLight® 800)
Antibodies
Anti-Fluorescein Antibody Dylight® 800 Conjugated is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: 1:10000-1:25000;
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: Fluorescein
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Goat
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty