Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFP Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 9F9.F9]

Supplier: Rockland Immunochemicals
600-305-215 600-305-215
RL600-305-215EA 1108 USD InStock
CA600-305-215 RL600-305-215
Anti-GFP Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 9F9.F9]
Antibodies
Monoclonal anti-GFP is designed to detect enhanced GFP and GFP containing recombinant Proteins. This antibody can be used to detect GFP by ELISA (sandwich or capture) for the direct binding of antigen.

Recommended Dilutions: ELISA: 1:2000 - 1:12000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Tag
Antigen: GFP
Clonality: Monoclonal
Clone: 9F9.F9
Conjugation: AP (Alkaline Phosphatase)
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty