Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-033 600-401-033
RL600-401-033EA 1290 USD InStock
RL600-401-033 CA600-401-033
Anti-HSA Rabbit Polyclonal Antibody
Antibodies
Anti-Human Serum Albumin Antibody is suitable for the detection of human serum albumin in blood and tissue samples. Antibody is suitable for western blotting to detect a single band of 67 kDa, the expected apparent molecular weight, and for use in ELISA.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Primary
Antigen: HSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty