Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COL2A1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-104S
RL600-401-104SEA 148 USD InStock
RL600-401-104S
Anti-COL2A1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:5000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:50 - 1:400; Immunoprecipitation: 1:100;

Type: Primary
Antigen: COL2A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty