Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COL4A1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-106S
RL600-401-106SEA 145 USD InStock
RL600-401-106S
Anti-COL4A1 Rabbit Polyclonal Antibody
Antibodies
Anti-Collagen antibodies have been used for indirect trapping ELISA for quantitation of antigen in serum using a standard curve, for immunoprecipitation and for native (non-denaturing, non-dissociating) PAGE and western blotting.

Recommended Dilutions: ELISA: 1:5000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:50 - 1:200; Immunoprecipitation: 1:100;

Type: Primary
Antigen: COL4A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty