Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TERT Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-252 600-401-252
RL600-401-252EA 534 USD InStock
RL600-401-252 CA600-401-252
Anti-TERT Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in immunoblotting, immunoprecipitation, and immunofluorescence microscopy.   In these assays, the antibody detects ectopically-expressed hTERT and high levels of endogenous hTERT.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Immunofluorescence: 1:500; Western Blot: 1:500- 1:2000; Immunochemistry: 1:500; Immunoprecipitation: 2 µl per mg of lysate;

Type: Primary
Antigen: TERT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty