Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-268 600-401-268
RL600-401-268EA 492 USD InStock
CA600-401-268 RL600-401-268
Anti-AKT1 Rabbit Polyclonal Antibody
Antibodies
This antibody is phospho specific for pS473 and is suitable for western blotting, immunochemistry (formalin-fixed paraffin-embedded sections), immunofluorescence and ELISA. By immunoblot a single band of the expected apparent molecular weight is obse

Recommended Dilutions: ELISA: 1:15000 - 1:60,000; Western Blot: 1:200 - 1:1000; Immunochemistry: 1:100 - 1:500;

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty