Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CREB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-270 600-401-270
RL600-401-270EA 518 USD InStock
RL600-401-270 CA600-401-270
Anti-CREB1 Rabbit Polyclonal Antibody
Antibodies
Anti-CREB pS133 antibody reacts with phosphorylated human CREB at pS133 and shows minimal reactivity by western blot with non-phosphorylated CREB and minimal reactivity (1%) by ELISA against the non-phosphorylated form of the immunizing peptide.

Recommended Dilutions: ELISA: 1:1000 - 1:6000; Western Blot: 1:500 - 1:2000; Immunochemistry: 20 µg/ml;

Type: Primary
Antigen: CREB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty