Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-272 600-401-272
RL600-401-272EA 1290 USD InStock
RL600-401-272 CA600-401-272
Anti-HP Rabbit Polyclonal Antibody
Antibodies
Human Haptoglobin Antibody is suitable for western blotting, ELISA and IHC. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:500,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Primary
Antigen: HP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty