Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPKAPK2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-281 600-401-281
RL600-401-281EA 518 USD InStock
RL600-401-281 CA600-401-281
Anti-MAPKAPK2 Rabbit Polyclonal Antibody
Antibodies
This antibody is suitable for western blotting, ELISA, immunoprecipitation and kinase assays.

Recommended Dilutions: ELISA: 1:15000; Western Blot: 1:2000;

Type: Primary
Antigen: MAPKAPK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty