Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RFP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-379 600-401-379
RL600-401-379EA 360 USD InStock
RL600-401-379 CA600-401-379
Anti-RFP Rabbit Polyclonal Antibody
Antibodies
Polyclonal anti-RFP is designed to detect RFP and its variants. This antibody can be used to detect RFP by ELISA (sandwich or capture) for the direct binding of antigen.

Recommended Dilutions: ELISA: 1:50000 - 1:100000; Immunofluorescence: 1:200-1:2000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:200 - 1:2000;

Type: Tag
Antigen: RFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: 0

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty