Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-6xHis Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-382S
RL600-401-382SEA 150 USD InStock
RL600-401-382S
Anti-6xHis Rabbit Polyclonal Antibody
Antibodies
Anti-6X His is optimally suited for monitoring expression of His-tagged fusion Proteins. As such, anti-6X His/6X His can be used to identify fusion Proteins that contain the 6X His epitope.

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:1000 - 1:3000; Immunochemistry: 1:500 - 1:2000;

Type: Tag
Antigen: 6xHis
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty