Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-412 600-401-412
RL600-401-412EA 518 USD InStock
RL600-401-412 CA600-401-412
Anti-CASP2 Rabbit Polyclonal Antibody
Antibodies
This affinity purified polyclonal antibody was tested by ELISA and immunoblotting and was found to be reactive with 48 kDa mouse and Rat caspase-2.

Recommended Dilutions: ELISA: 1:1500 to 1:6000; Western Blot: 1:750;

Type: Primary
Antigen: CASP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty