Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NF2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-414 600-401-414
RL600-401-414EA 518 USD InStock
RL600-401-414 CA600-401-414
Anti-NF2 Rabbit Polyclonal Antibody
Antibodies
This phospho-specific polyclonal antibody was tested by immunoblotting and ELISA.  By ELISA, the antibody was found to be reactive with the phosphorylated form of the immunizing peptide and minimally reactive with the non-phosphorylated form of the immuni

Recommended Dilutions: ELISA: 1:1200,000; Western Blot: 1:500- 1:2000;

Type: Primary
Antigen: NF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty