Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PPARA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-421S
RL600-401-421SEA 148 USD InStock
RL600-401-421S
Anti-PPARA Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and by western blot. Expect a single band approximately 52 kDa in size corresponding to PPAR alpha by western blot in the appropriate tissue or cell lysate.

Recommended Dilutions: ELISA: 1:8000 - 1:32000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:100-1:300;

Type: Primary
Antigen: PPARA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Bovine, Dog, Golden Hamster, Boar, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty