Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MTOR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-422 600-401-422
RL600-401-422EA 518 USD InStock
CA600-401-422 RL600-401-422
Anti-MTOR Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in immunochemistry, ELISA and western blotting. Western blotting shows reactivity specific for phospho mTOR detecting a band at approximately 250 kDa.

Recommended Dilutions: ELISA: 1:10000 - 1:100000; Western Blot: 1:500 - 1:2000; Immunochemistry: 5.0 µg/ml;

Type: Primary
Antigen: MTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty