Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CBX4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-478 600-401-478
RL600-401-478EA 518 USD InStock
CARL600401-478 RL600-401-478
Anti-CBX4 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in immunofluorescence staining of cultured cells and western blotting using transfected cell lysates.  The antibody is also functional by ELISA.  Reactivity in other immunoassays is unknown.

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Immunofluorescence: 1:100 - 1:400; Western Blot: 1:1000 - 1:4000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CBX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty