Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AURKB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-677S
RL600-401-677SEA 145 USD InStock
RL600-401-677S
Anti-AURKB Rabbit Polyclonal Antibody
Antibodies
Phospho pT232 Aurora B Antibody is affinity-purified antibody has been tested for use in ELISA and by western blot. See below for specific protocol. Expect a band approximately 39 kDa in size corresponding to Aurora Kinase B by western blotting.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:250 - 1:2000;

Type: Primary
Antigen: AURKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty