Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SPRY4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-697 600-401-697
RL600-401-697EA 518 USD InStock
CARL600401-697 RL600-401-697
Anti-SPRY4 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA, immunochemistry and western blot.  Specific conditions for reactivity should be optimized by the end user.Expect a band (maybe a doublet) ~35 kDa in size.

Recommended Dilutions: ELISA: 1:3000 - 1:12000; Western Blot: 1:100 - 1:1000; Immunochemistry: 1 µg/ml to 10 µg/ml;

Type: Primary
Antigen: SPRY4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty