Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACTB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-886 600-401-886
RL600-401-886EA 436 USD InStock
RL600-401-886 CA600-401-886
Anti-ACTB Rabbit Polyclonal Antibody
Antibodies
Anti-beta Actin Antibody has been tested for use in ELISA,.

  • Recommended Dilutions: ELISA: 1:10000 - 1:40000; Immunofluorescence: 1:500 - 1:2000; Western Blot: 1:1000 - 1:4000;

Type: Primary
Antigen: ACTB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Goat, Guinea Pig, Hamster, Human, Monkey, Mouse, Rat, Sheep, Pig

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty