Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MTOR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-897 600-401-897
RL600-401-897EA 518 USD InStock
RL600-401-897 CA600-401-897
Anti-MTOR Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody is suitable for use in ELISA and western blotting. ELISA data demonstrate reactivity against both phosphorylated and non-phosphorylated mTOR at S2448 and western blotting shows a band at approximately 250 kDa.

Recommended Dilutions: ELISA: 1:50000 - 1:100000; Western Blot: 1:250 - 1:2000;

Type: Primary
Antigen: MTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty