Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NOXO1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-899 600-401-899
RL600-401-899EA 518 USD InStock
RL600-401-899 CA600-401-899
Anti-NOXO1 Rabbit Polyclonal Antibody
Antibodies
Anti-NOXO1 antibody has been tested for use in ELISA, immunochemistry and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 50 kDa in size corresponding to NOXO1 Protein by western blot.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 5-10 µg/ml;

Type: Primary
Antigen: NOXO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty