Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AcK Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-939 600-401-939
RL600-401-939EA 518 USD InStock
RL600-401-939 CARL600401-939
Anti-AcK Rabbit Polyclonal Antibody
Antibodies
Anti-Lysine Acetylated Antibody is suitable for use in ELISA, western blotting, immunofluorescence microscopy, and immunoprecipitation assays. Although not tested, this antibody is likely functional in RIA, flow cytometry, and immunochemistry.

Recommended Dilutions: ELISA: 1:1000-1:2500; Western Blot: 1:500-1:1000; Immunoprecipitation: 10 µg/mg;

Type: Primary
Antigen: AcK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Avian, Mammalian

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty