Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AUB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-947 600-401-947
RL600-401-947EA 518 USD InStock
RL600-401-947 CARL600401-947
Anti-AUB Rabbit Polyclonal Antibody
Antibodies
Aubergine affinity-purified antibody has been tested for use in ELISA.  Specific conditions for reactivity should be optimized by the end user. By western blot a band approximately 98 kDa in size.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:1000 - 1:10000

Type: Primary
Antigen: AUB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila Melanogaster

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty