Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IRAK3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-954 600-401-954
RL600-401-954EA 518 USD InStock
RL600-401-954 CARL600401-954
Anti-IRAK3 Rabbit Polyclonal Antibody
Antibodies
Anti-IRAK-M Antibody is suitable for use in ELISA, immunochemistry and western blot.  Expect a band approximately 68 kDa in size corresponding to IRAK-M Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:300 - 1:1000;

Type: Primary
Antigen: IRAK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty