Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CENPU Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-A91S 600-401-A91S
RL600-401-A91SEA 148 USD InStock
CA600-401-A91S RL600-401-A91S
Anti-CENPU Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CENPU
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty