Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GDF15 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-B07 600-401-B07
RL600-401-B07EA 518 USD InStock
RL600-401-B07 CA600-401-B07
Anti-GDF15 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody is suitable for ELISA and western blotting of human and mouse NAG-1 Protein.

Recommended Dilutions: ELISA: 1:300,000; Western Blot: 1:2000 - 1:10000;

Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty